1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-{[(tert-Butoxycarbonyl)amino]methyl}-3-azabicyclo[3.1.0]hexane, tert-Butyl (3-azabicyclo[3.1.0]hexan-1-ylmethyl)carbamate, tert-Butyl [(3-azabicyclo[3.1.0]hex-1-yl)methyl]carbamate

OR16003-1G
APOSOR16003-1GEA 337 EUR
APOSOR16003-1G
1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected
1-(Aminomethyl)-3-azabicyclo[3.1.0]hexane, 1-BOC protected
Formel: C₁₁H₂₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11035741
CAS-Nummer: 134574-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR