2-Methylpiperazine, N1-CBZ protected 96%
Lieferant: Apollo Scientific
Synonyme:
Benzyl 2-methylpiperazine-1-carboxylate
OR16019-1G
APOSOR16019-1GEA
261
EUR
APOSOR16019-1G
2-Methylpiperazine, N1-CBZ protected 96%
2-Methylpiperazine, N1-CBZ protected
Formel:
C₁₃H₁₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115058 CAS-Nummer: 444666-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...