4-Nitrophenylglyoxal hydrate 95%

Lieferant: Apollo Scientific
Gefahr

OR1604-10G
APOSOR1604-10GEA 243 EUR
APOSOR1604-10G
4-Nitrophenylglyoxal hydrate 95%
4-Nitrophenylglyoxal hydrate
Formel: C₈H₅NO₄·H₂O
Molecular Weight: 197.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776822
CAS-Nummer: 4996-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR