(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid

Lieferant: Apollo Scientific

OR16060-1G OR16060-5G
APOSOR16060-1GEA 538 EUR
APOSOR16060-1G APOSOR16060-5G
(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid
(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid
Formel: C₁₀H₁₂O₄
Molecular Weight: 196.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00267156
CAS-Nummer: 34487-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR