1-Pyrimidin-2-ylethanone 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR16083-1G
APOSOR16083-1GEA
24.8
EUR
APOSOR16083-1G
1-Pyrimidin-2-ylethanone 97%
1-Pyrimidin-2-ylethanone
Formel:
C₆H₆N₂O Schmelzpunkt: 52…53 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08361636 CAS-Nummer: 53342-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...