3-Aminothiophene-2-carboxylic acid potassium salt
Lieferant: Apollo Scientific
OR16085-1G
APOSOR16085-1GEA
306
EUR
APOSOR16085-1G
3-Aminothiophene-2-carboxylic acid potassium salt
3-Aminothiophene-2-carboxylic acid potassium salt
Formel:
C₅H₄NO₂S·K Lagertemperatur: Raumtemperatur |
MDL:
MFCD11227267 CAS-Nummer: 1210469-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...