4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR16087-1G
APOSOR16087-1GEA 21.9 EUR
APOSOR16087-1G
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid
Formel: C₁₇H₁₄N₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11521324
CAS-Nummer: 641569-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR