4-(Dimethylamino)phenylglyoxal hydrate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N,N-Dimethyl-4-(oxoacetyl)aniline hydrate, [4-(Dimethylamino)phenyl](oxo)acetaldehyde hydrate

OR1612-150MG
APOSOR1612-150MGEA 65.4 EUR
APOSOR1612-150MG
4-(Dimethylamino)phenylglyoxal hydrate 95%
4-(Dimethylamino)phenylglyoxal hydrate
Formel: C₁₀H₁₁NO₂·H₂O
Molecular Weight: 195.215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234649
CAS-Nummer: 1171790-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR