(3-Amino-4-hydroxyphenyl)acetic acid

Lieferant: Apollo Scientific
Achtung

OR16144-250MG
APOSOR16144-250MGEA 221 EUR
APOSOR16144-250MG
(3-Amino-4-hydroxyphenyl)acetic acid
(3-Amino-4-hydroxyphenyl)acetic acid
Formel: C₈H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04125028
CAS-Nummer: 38196-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR