2-Ethylbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

OR16146-100G OR16146-25G OR16146-5G
APOSOR16146-100GEA 149 EUR
APOSOR16146-100G APOSOR16146-25G APOSOR16146-5G
2-Ethylbenzoesäure 98%
2-Ethylbenzoic acid
Formel: C₉H₁₀O₂
Molecular Weight: 150.1700 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045838
CAS-Nummer: 612-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR