2-Ethylbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR16146-100G
OR16146-25G
OR16146-5G
APOSOR16146-100GEA
149
EUR
APOSOR16146-100G
APOSOR16146-25G
APOSOR16146-5G
2-Ethylbenzoesäure 98%
2-Ethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150.1700 g/mol Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045838 CAS-Nummer: 612-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...