2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid

Lieferant: Apollo Scientific

OR16148-1G
APOSOR16148-1GEA 0 EUR
APOSOR16148-1G
2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid
2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid
Formel: C₁₁H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00022590
CAS-Nummer: 80022-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR