6-Amino-3-brom-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR16151-1G
OR16151-5G
APOSOR16151-1GEA
190
EUR
APOSOR16151-1G
APOSOR16151-5G
6-Amino-3-brom-1H-indazol
6-Amino-3-bromo-1H-indazole
Formel:
C₇H₆BrN₃ Schmelzpunkt: 220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781649 CAS-Nummer: 52347-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...