5-Nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Lieferant: Apollo Scientific
|
Achtung
|
OR16152-1G
APOSOR16152-1GEA
132
EUR
APOSOR16152-1G
5-Nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
5-Nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Formel:
C₈H₆N₂O₄ Molecular Weight: 194,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100144 CAS-Nummer: 132522-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...