3-(3-Nitrophenyl)thiophene

Lieferant: Apollo Scientific

Synonyme: 3-(Thien-3-yl)nitrobenzene

OR16154-1G
APOSOR16154-1GEA 0 EUR
APOSOR16154-1G
3-(3-Nitrophenyl)thiophene
3-(3-Nitrophenyl)thiophene
Formel: C₁₀H₇NO₂S
Molecular Weight: 205.233 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11109509
CAS-Nummer: 30011-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR