N-(2-Chloroethyl)-1H-pyrazole-1-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[(2-Chloroethyl)carbamoyl]-1H-pyrazole, 1-{[(2-Chloroethyl)amino]carbonyl}-1H-pyrazole

OR16157-1G
APOSOR16157-1GEA 60.9 EUR
APOSOR16157-1G
N-(2-Chloroethyl)-1H-pyrazole-1-carboxamide
N-(2-Chloroethyl)-1H-pyrazole-1-carboxamide
Formel: C₆H₈ClN₃O
Molecular Weight: 173,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109511
CAS-Nummer: 1209343-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR