1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
Lieferant: Apollo Scientific
Synonyme:
1-(2,3-Dihydro-7-nitro-1,1,3,3,6-pentamethyl-1H-inden-5-yl)imidazolidin-2-one
OR16160-1G
APOSOR16160-1GEA
232
EUR
APOSOR16160-1G
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)imidazolidin-2-one
Formel:
C₁₇H₂₃N₃O₃ Molecular Weight: 317,383 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109514 CAS-Nummer: 1209101-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...