7-Chloro-8-methylquinoline

Lieferant: Apollo Scientific

OR16164-1G
APOSOR16164-1GEA 0 EUR
APOSOR16164-1G
7-Chloro-8-methylquinoline
7-Chloro-8-methylquinoline
Formel: C₁₀H₈ClN
Schmelzpunkt: 45…47 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09743893
CAS-Nummer: 78941-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR