3-Isoxazolcarbonsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Isoxazole-3-carboxylic Acid

OR16167-25G
APOSOR16167-25GEA 126 EUR
APOSOR16167-25G
3-Isoxazolcarbonsäure 96%
3-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113.07 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738796
CAS-Nummer: 3209-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR