3-Chlorophenylglyoxal hydrate 95%

Lieferant: Apollo Scientific
Achtung

OR1625-250MG
APOSOR1625-250MGEA 0 EUR
APOSOR1625-250MG
3-Chlorophenylglyoxal hydrate 95%
3-Chlorophenylglyoxal hydrate
Formel: C₈H₅ClO₂·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08705874
CAS-Nummer: 177288-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR