(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen, (2R)-2-(6-Methoxynaphth-2-yl)propionic acid, R-Naproxen, 2-[(1R)-1-Carboxyethyl]-6-methoxynaphthalene

OR16409-5G
APOSOR16409-5GEA 121 EUR
APOSOR16409-5G
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid
(2R)-2-(6-Methoxynaphth-2-yl)propanoic acid
Formel: C₁₄H₁₄O₃
Molecular Weight: 230.259 g/mol
Schmelzpunkt: 156…158 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00870716
CAS-Nummer: 23979-41-1
EINECS: 245-966-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR