3-Nitroisonicotinic acid

Lieferant: Apollo Scientific
Achtung

OR16446-1G
APOSOR16446-1GEA 0 EUR
APOSOR16446-1G
3-Nitroisonicotinic acid
3-Nitroisonicotinic acid
Formel: C6H4N2O4
Lagertemperatur: Raumtemperatur
MDL: MFCD04114244
CAS-Nummer: 59290-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR