2,6-Dimethylnicotinic acid 98%

Lieferant: Apollo Scientific
Achtung

OR16447-1G
APOSOR16447-1GEA 71.2 EUR
APOSOR16447-1G
2,6-Dimethylnicotinic acid 98%
2,6-Dimethylnicotinic acid
Formel: C₈H₉NO₂
Schmelzpunkt: 168…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738677
CAS-Nummer: 5860-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR