4-(Piperidin-1-yl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR1646-1G
APOSOR1646-1GEA
21.9
EUR
APOSOR1646-1G
4-(Piperidin-1-yl)benzaldehyde 95%
4-(Piperidin-1-yl)benzaldehyde
Formel:
C₁₂H₁₅NO Molecular Weight: 189,2500 g/mol Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00778395 CAS-Nummer: 10338-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...