5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR16462-500MG
APOSOR16462-500MGEA
116
EUR
APOSOR16462-500MG
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
Formel:
C₉H₁₃N₃ Molecular Weight: 163.22 g/mol Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846194 CAS-Nummer: 269726-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...