Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carboxylate

OR16468-250MG
APOSOR16468-250MGEA 34.8 EUR
APOSOR16468-250MG
Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,2 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11848180
CAS-Nummer: 142166-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR