Dibromoisocyanuric acid

Lieferant: Apollo Scientific
Gefahr

OR16500-25G OR16500-5G OR16500-1G
APOSOR16500-25GEA 244 EUR
APOSOR16500-25G APOSOR16500-5G APOSOR16500-1G
Dibromoisocyanuric acid
Dibromoisocyanuric acid
Formel: C₃HBr₂N₃O₃
Schmelzpunkt: 309 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00463941
CAS-Nummer: 15114-43-9
UN: 3085
ADR: 5.1+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR