1-Tritylimidazol-2-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Trityl-1H-imidazole-2-carbaldehyde

OR1652-250MG
APOSOR1652-250MGEA 34.8 EUR
APOSOR1652-250MG
1-Tritylimidazol-2-carbaldehyd
1-Tritylimidazol-2-carbaldehyd
Formel: C₂₃H₁₈N₂O
Molecular Weight: 338.400 g/mol
Schmelzpunkt: 189…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453110
CAS-Nummer: 67478-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR