5-[(Morpholin-4-yl)methyl]thiophene-3-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Gefahr

OR16540-250MG
APOSOR16540-250MGEA 33.5 EUR
APOSOR16540-250MG
5-[(Morpholin-4-yl)methyl]thiophene-3-boronic acid, pinacol ester
5-[(Morpholin-4-yl)methyl]thiophene-3-boronic acid, pinacol ester
Formel: C₁₅H₂₄BNO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD11841079
CAS-Nummer: 364794-85-4
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR