4-(4-Bromo-1,3-thiazol-2-yl)morpholine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-Bromothiazol-2-yl)morpholine, 4-Bromo-2-(morpholin-4-yl)-1,3-thiazole
OR16542-250MG
APOSOR16542-250MGEA
42.2
EUR
APOSOR16542-250MG
4-(4-Bromo-1,3-thiazol-2-yl)morpholine 97%
4-(4-Bromo-1,3-thiazol-2-yl)morpholine
Formel:
C₇H₉BrN₂OS Molecular Weight: 249,128 g/mol Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09607696 CAS-Nummer: 1017781-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...