5-Methyl-2-pyrazinamin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-methylpyrazine
OR16560-1G
APOSOR16560-1GEA
31.9
EUR
APOSOR16560-1G
5-Methyl-2-pyrazinamin
5-Methylpyrazin-2-amine
Formel:
C5H7N3 Molecular Weight: 109,13 g/mol Schmelzpunkt: 111…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437664 CAS-Nummer: 5521-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...