Tetrahydrofuran-3-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR16566-1G
OR16566-250MG
OR16566-100MG
APOSOR16566-1GEA
301
EUR
APOSOR16566-1G
APOSOR16566-250MG
APOSOR16566-100MG
Tetrahydrofuran-3-carbonyl chloride
Tetrahydrofuran-3-carbonyl chloride
Formel:
C₅H₇ClO₂ Siedepunkt: 75…76 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656696 CAS-Nummer: 69595-02-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...