Isoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR16580-25G
APOSOR16580-25GEA 219 EUR
APOSOR16580-25G
Isoquinoline-3-carboxylic acid
Isoquinoline-3-carboxylic acid
Formel: C₁₀H₇NO₂
Molecular Weight: 173,1681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075137
CAS-Nummer: 6624-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR