Benzothiazol-2-ol 98%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxybenzothiazole
OR16606-100G
APOSOR16606-100GEA
97.3
EUR
APOSOR16606-100G
Benzothiazol-2-ol 98%
Benzothiazol-2-ol
Formel:
C₇H₅NOS Molecular Weight: 151,18 g/mol Schmelzpunkt: 137…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022868 CAS-Nummer: 934-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...