(2-Benzyloxyphenyl)essigsäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR1661-1G
APOSOR1661-1GEA
0
EUR
APOSOR1661-1G
(2-Benzyloxyphenyl)essigsäure
(2-Benzyloxyphenyl)essigsäure
Formel:
C₆H₅CH₂OC₆H₄CH₂COOH Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016822 CAS-Nummer: 22047-88-7 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...