1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(4-Methylpiperazin-1-yl)pyridine-5-boronic acid pinacol ester, [6-(4-Methylpiperazin-1-yl)pyridin-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Methyl-4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine, 6-(4-Methylpiperazin-1-yl)pyridine-3-boronic acid Pinacol ester
OR16630-250MG
APOSOR16630-250MGEA
21.9
EUR
APOSOR16630-250MG
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 98%
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine
Formel:
C₁₆H₂₆BN₃O₂ Schmelzpunkt: 125…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437995 CAS-Nummer: 918524-63-7 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...