4-Boronobenzolsulfonamidpinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide, 4-Sulphamoylbenzeneboronic acid, pinacol ester

OR16633-500MG
APOSOR16633-500MGEA 72.5 EUR
APOSOR16633-500MG
4-Boronobenzolsulfonamidpinakolester 98%
4-Boronobenzolsulfonamidpinakolester
Formel: C₁₂H₁₈BNO₄S
Molecular Weight: 283,1516 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06657806
CAS-Nummer: 214360-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR