4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methyl-1-piperazinyl)phenylboronic Acid Pinacol Ester

OR16634-5G
APOSOR16634-5GEA 60.9 EUR
APOSOR16634-5G
4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester
4-(4-Methyl-1-piperazinyl)benzeneboronic acid pinacol ester
Formel: C₁₇H₂₇BN₂O₂
Molecular Weight: 302.2195 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795650
CAS-Nummer: 747413-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR