5-Methyl-1H-indole-3-carbaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR1664-250MG
APOSOR1664-250MGEA
0
EUR
APOSOR1664-250MG
5-Methyl-1H-indole-3-carbaldehyde 98%
5-Methyl-1H-indole-3-carbaldehyde
Formel:
C10H9NO Schmelzpunkt: 148…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022720 CAS-Nummer: 52562-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...