5-Bromobenzo[c]isothiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2,1-benzothiazole

OR16642-100MG
APOSOR16642-100MGEA 174 EUR
APOSOR16642-100MG
5-Bromobenzo[c]isothiazole
5-Bromobenzo[c]isothiazole
Formel: C₇H₄BrNS
Molecular Weight: 214,082 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08706414
CAS-Nummer: 20712-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR