6-Bromobenzo[c]isothiazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-2,1-benzothiazole

OR16643-250MG
APOSOR16643-250MGEA 603 EUR
APOSOR16643-250MG
6-Bromobenzo[c]isothiazole 95%
6-Bromobenzo[c]isothiazole
Formel: C₇H₄BrNS
Molecular Weight: 214,082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176692
CAS-Nummer: 20712-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR