3-Bromo-2-methylbenzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3-(bromomethyl)-2-methylbenzene, α,3-Dibromo-o-xylene
OR16663-1G
OR16663-250MG
APOSOR16663-1GEA
230
EUR
APOSOR16663-1G
APOSOR16663-250MG
3-Bromo-2-methylbenzyl bromide 97%
3-Bromo-2-methylbenzyl bromide
Formel:
C₈H₈Br₂ Schmelzpunkt: 31…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847532 CAS-Nummer: 112299-62-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...