3-Aminopentanoic acid

Lieferant: Apollo Scientific
Achtung

OR16680-1G OR16680-5G OR16680-10G
APOSOR16680-1GEA 89.9 EUR
APOSOR16680-1G APOSOR16680-5G APOSOR16680-10G
3-Aminopentanoic acid
3-Aminopentanoic acid
Formel: C5H11NO2
Lagertemperatur: Raumtemperatur
CAS-Nummer: 18664-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR