1-Methylcyclopropanamine hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

OR16691-5G
APOSOR16691-5GEA 75.4 EUR
APOSOR16691-5G
1-Methylcyclopropanamine hydrochloride 98%
1-Methylcyclopropanamine hydrochloride
Formel: C₄H₁₀ClN
Molecular Weight: 107.5800 g/mol
Schmelzpunkt: 196…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11109576
CAS-Nummer: 88887-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR