7-Nitroindole-3-carboxyaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR1670-1G
APOSOR1670-1GEA 21.9 EUR
APOSOR1670-1G
7-Nitroindole-3-carboxyaldehyde 95+%
7-Nitroindole-3-carboxyaldehyde
Formel: C₉H₆N₂O₃
Molecular Weight: 190,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01074519
CAS-Nummer: 10553-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR