4-Oxa-1,9-diazaspiro[5.5]undecane, N9-BOC protected

Lieferant: Apollo Scientific
Achtung

OR16701-250MG
APOSOR16701-250MGEA 0 EUR
APOSOR16701-250MG
4-Oxa-1,9-diazaspiro[5.5]undecane, N9-BOC protected
4-Oxa-1,9-diazaspiro[5.5]undecane, N9-BOC protected
Formel: C₁₃H₂₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12198543
CAS-Nummer: 1160247-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR