1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Oxo-1-oxa-7-azaspiro[4.5]decane hydrochloride
OR16705-250MG
APOSOR16705-250MGEA
197
EUR
APOSOR16705-250MG
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
1-Oxa-7-azaspiro[4.5]decan-2-one hydrochloride
Formel:
C₈H₁₃NO₂·ClH Molecular Weight: 191,655 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18642911 CAS-Nummer: 1314961-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...