6-Oxa-1-azaspiro[3.3]heptane oxalate

Lieferant: Apollo Scientific
Achtung

OR16706-250MG
APOSOR16706-250MGEA 374 EUR
APOSOR16706-250MG
6-Oxa-1-azaspiro[3.3]heptane oxalate
6-Oxa-1-azaspiro[3.3]heptane oxalate
Formel: C₇H₁₁NO₅
Lagertemperatur: Kühlschrank
MDL: MFCD21364487
CAS-Nummer: 1359655-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR