tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Benzyloxy)-3-formyl-1H-indole, N-BOC protected
OR1674-250MG
APOSOR1674-250MGEA
39.3
EUR
APOSOR1674-250MG
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate
Formel:
C₂₁H₂₁NO₄ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
914348-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...