6-(Benzyloxy)-1H-indol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Benzyloxyindole

OR1675-1G
APOSOR1675-1GEA 21.9 EUR
APOSOR1675-1G
6-(Benzyloxy)-1H-indol 98%
6-(Benzyloxy)-1H-indole
Formel: C15H13NO
Molecular Weight: 223,28 g/mol
Schmelzpunkt: 114…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00053554
CAS-Nummer: 15903-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR